Anti-H3F3A Rabbit Polyclonal Antibody
Catalog # ROCK600-401-P10
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:H3 histone, family 3A
- Antigen symbol:H3F3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Primates,Drosophila,Plant,Xenopus,C. elegans,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:3020
- Antigen synonyms:H3.3A|H3F3
- Modification:monomethyl-Lys79
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide, 30% Glycerol
- Storage temperature:Cool
- Concentration:0,50 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Histone H3 K79-Me1 affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide surrounding the Lys79 site of human Histone H3.
- Purification:affinity-purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Anti-Histone H3 K79-Me1 antibody is has been tested for use in Western Blot, ChIP, Immunofluorescence, and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15kDa.
Recommended Dilutions: ChIP: 2 µg; Immunofluorescence: 1:50 - 1:100; Western Blot: 0.5µg/ml
Type: Primary
Antigen: H3F3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primate, C. elegans, Drosophila, Plant, Rat, Xenopus