Anti-TCRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: H57-597]
Catalog # ROCK200-B02-N92
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:T cell receptor beta chain
- Antigen symbol:TCRB
- Clonality:Monoclonal
- Clone:H57-597
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Armenian hamster
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:21577
- Antigen synonyms:TCRbeta|Tib
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-TCRbeta Antibody (Monoclonal) was produced by repeated immunizations with TCRbeta antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Anti-TCRbeta is useful for Immunofluorescence, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!
Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse