Order Entry
Ireland
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: RA3-6B2]
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: RA3-6B2]
Catalog # ROCK200-502-N84
undefined
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: RA3-6B2]
Catalog # ROCK200-502-N84

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD45R
  • Clonality:
    Monoclonal
  • Clone:
    RA3-6B2
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD45R (B220) Antibody (Monoclonal) was produced by repeated immunizations with CD45R antigen.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Anti-CD45R (B220) is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse