Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI30]
Catalog # ROCK200-326-N68
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:HI30
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|LCA
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,025 mg/ml Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD45 Antibody (Monoclonal) was produced by repeated immunizations with CD45 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Anti-CD45 is useful for Flow Cytometry, immunochemistry, and Western Blotting. Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human