IgG1 Isotype Control (Biotin)
Catalog # ROCK012-006-040
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Drosophila,Plant,Xenopus,Chicken,C. elegans,Human,Rat,Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Rat IgG1 isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.
Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!
Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: