Order Entry
Ireland
ContactUsLinkComponent
IgG2a kappa Isotype Control (Unconjugated)
IgG2a kappa Isotype Control (Unconjugated)
Catalog # ROCK012-001-332
IgG2a kappa Isotype Control (Unconjugated)
Catalog # ROCK012-001-332

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Rat IgG2a Kappa isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: RG2aK
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: