IgG1 lambda Isotype Control (Unconjugated)
Catalog # ROCK012-001-331
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 lambda
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Rat IgG1 Lambda isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.
Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;
Type: Control
Antigen: IgG1 λ
Clonality: Monoclonal
Clone: RG1L
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: