Order Entry
Ireland
ContactUsLinkComponent
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Peroxidase)
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Peroxidase)
Catalog # ROCK712-1331
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Peroxidase)
Catalog # ROCK712-1331

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG F(ab')2
  • Clonality:
    Polyclonal
  • Conjugation:
    Peroxidase
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    Bovine, Horse, Human
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,25 M Sodium Chloride, pH 7,2, 15 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    0,8 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-Rat IgG F(ab')2 fragment was produced by repeated immunization with Rat IgG F(ab')2 fragment in goat.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    500 µl
  • Pk:
    500 µG

Specifications

About this item

Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rat