Order Entry
Ireland
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-IgG (H+L) Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ROCK709-1816
undefined
Anti-IgG (H+L) Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ROCK709-1816

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    Bovine, Horse, Mouse, Rat
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml by absorbance = 82,0 at 565 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-Human IgG (H&L) was produced by repeated immunization with human IgG whole molecule in goat.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Human F(ab')2 IgG (H&L) phycoerythrin conjugated antibody is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity., lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human