Order Entry
Ireland
ContactUsLinkComponent
 
Anti-IgG (H+L) Guniea pig Polyclonal Antibody (Biotin)
  ROCK611-206-122
undefined
Anti-IgG (H+L) Guniea pig Polyclonal Antibody (Biotin)
  ROCK611-206-122

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Guinea pig
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Rabbit
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    Human, Goat, Mouse
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Rabbit IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Guinea Pig
Isotype:
Reactivity: Rabbit