Anti-NEU2 Rabbit Polyclonal Antibody
ROCK600-401-850
- Antibody type:Primary
- Antigen name:Neuraminidase 2
- Antigen symbol:NEU2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:4759
- Antigen synonyms:SIAL2
- Amino acid number:110 to 124
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,9 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 110-124 of Human Neu2.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
This antibody is suitable for western blotting,.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: NEU2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human