Anti-ATM Rabbit Polyclonal Antibody
ROCK600-401-398
- Antibody type:Primary
- Antigen name:ATM serine/threonine kinase
- Antigen symbol:ATM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:472
- Antigen synonyms:AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
- Amino acid number:1974 to 1988
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Sequence:SLAFEEGSQSTTISS
- Storage temperature:Cool
- Concentration:1,1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This antibody was affinity purified from whole rabbit serum prepared by repeated immunizations with a synthetic peptide S-L-A-F-E-E-G-S-Q-S-T-T-I-S-S corresponding to aa 1974-1988 of human ATM conjugated to KLH using maleimide.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Affinity purified rabbit anti-ATM has been tested by ELISA, immunofluorescence microscopy, western blotting and biological assays against both the native and recombinant forms of the Protein.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;
Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse