Anti-GFP Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9F9.F9]
Catalog # ROCK600-303-215
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Monoclonal
- Clone:9F9.F9
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Multiple
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Thimerosal, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-Green Fluorescent Protein (GFP) is produced by immunizing GFP containing fusion Protein that corresponds to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
- Purification:affinity-purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone: 9F9.F9
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan