Anti-ATM Mouse Monoclonal Antibody (Unconjugated) [clone: 7C10D8]
ROCK200-301-500
- Antibody type:Primary
- Antigen name:ATM serine/threonine kinase
- Antigen symbol:ATM
- Clonality:Monoclonal
- Clone:7C10D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:472
- Antigen synonyms:AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
- Modification:phospho-Ser1981
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This antibody was produced from a synthetic peptide S-L-A-F-E-E-G-Sp-Q-S-T-T-I-S-S corresponding to aa 1974-1988 of human ATM.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
This Anti-ATM Protein Kinase pS1981 (MOUSE) Monoclonal Antibody clone has been optimized for IHC, but may also be used for western blotting, flow cytometry, immunoprecipitation, or immunofluorescence microscopy.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 1:100 - 1:500;
Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 7C10D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse