Anti-Bilirubin Oxidase Goat Polyclonal Antibody
ROCK100-101-212
- Antibody type:Primary
- Antigen name:Bilirubin Oxidase (Myrothecium verrucaria)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Myrothecium verrucaria
- Western blot:Yes
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:95 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Bilirubin Oxidase (Myrothecium verrucaria)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Primary Goat Anti-BILIRUBIN OXIDASE (Myrothecium verrucaria) Reacts with Pan
Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: Bilirubin Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Myrothecium Verrucaria