Order Entry
Ireland
ContactUsLinkComponent
Anti-c-Raf (pTyr 340 , Tyr 341 ) Rabbit Polyclonal Antibody
Anti-c-Raf (pTyr 340 , Tyr 341 ) Rabbit Polyclonal Antibody
  553009-10
Anti-c-Raf (pTyr 340 , Tyr 341 ) Rabbit Polyclonal Antibody
  553009-10

 

  • Antibody type:
    Primary
  • Antigen symbol:
    c-Raf
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Modification:
    phospho-Tyr 340/341
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    10 Tests
  • Pk:
    10 Tests

 

 

Type: Primary
Antigen: c-Raf (pTyr 340 , Tyr 341 )
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: