Order Entry
Ireland
ContactUsLinkComponent
Anti-PP2A/Balpha (14-27) (PR55) Rabbit Polyclonal Antibody
Anti-PP2A/Balpha (14-27) (PR55) Rabbit Polyclonal Antibody
  539521-100
Anti-PP2A/Balpha (14-27) (PR55) Rabbit Polyclonal Antibody
  539521-100

 

  • Antibody type:
    Primary
  • Antigen name:
    PP2A/B alpha
  • Antigen symbol:
    PP2A/B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: PP2A/Balpha (14-27) (PR55)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: