Anti-PRICKLE3 / LMO6 Goat Polyclonal Antibody
EVBIEB08130
- Antibody type:Primary
- Antigen symbol:PRICKLE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:LMO6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PRICKLE3 / LMO6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat