Anti-ENPP1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB06948
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:ectonucleotide pyrophosphatase/phosphodiesterase 1
- Antigen symbol:ENPP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat