Order Entry
Ireland
ContactUsLinkComponent
Anti-SERCA2 ATPase Rabbit Polyclonal Antibody
Anti-SERCA2 ATPase Rabbit Polyclonal Antibody
  BSBTPA1720
undefined
Anti-SERCA2 ATPase Rabbit Polyclonal Antibody
  BSBTPA1720

 

  • Antibody type:
    Primary
  • Antigen name:
    Sarcoplasmic or Endoplasmic Reticulum Calcium ATPase 2
  • Antigen symbol:
    SERCA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    Sarcoplasmic or Endoplasmic Reticulum Calcium ATPase 2|ATP2B|DAR|DD|SERCA2
  • Amino acid number:
    665 to 679
  • Sequence:
    QRDACLNARCFARVE
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human SERCA2 ATPase (665-679aa QRDACLNARCFARVE), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Sarcoplasmic/endoplasmic reticulum calcium ATPase 2(ATP2A2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SERCA2 ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat