- Antibody type:Primary
- Antigen name:Receptor tyrosine-protein kinase erbB-2
- Antigen symbol:HER2
- Clonality:Monoclonal
- Clone:e2-4001 + 3B5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:2064
- Antigen synonyms:herstatin|NEU|MLN19|Tyrosine kinase-type cell surface receptor HER2|c-erb B2/neu protein|MLN 19|CD340|Metastatic lymph node gene 19 protein|p185erbB2|Receptor tyrosine-protein kinase erbB-2|Proto-oncogene c-ErbB-2|Proto-oncogene Neu|metas|NGL
- Storage buffer:PBS, 0.2% BSA, 0.09% sodium azide, pH 7.4
- Storage temperature:4° C
- Concentration:0.1 mg/ml
- Shipping temperature:Ice
- Immunogen:Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5)
- Purification:Protein G
- Size:500 µL
- Pk:500 µl
MA5-14057 targets HER-2 in IHC (P), IP, ICC/IF, FACS and WB applications and shows reactivity with Human samples. The MA5-14057 immunogen is cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5). This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
Type: Primary
Antigen: HER2
Clonality: Monoclonal
Clone: e2-4001 + 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human