- Antibody type:Primary
- Antigen name:Sodium/Calcium Exchanger
- Antigen symbol:SLC
- Clonality:Monoclonal
- Clone:C2C12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Rabbit,Human,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:Liquid
- Gene ID:100135620, 475738, 6546, 100328570, 29715, 20541
- Antigen synonyms:NKX|Na+/C2+ exchanger
- Storage buffer:ascites, 0.05% sodium azide
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Shipping temperature:Ice
- Immunogen:Purified canine cardiac sodium/calcium exchanger.
- Size:100 µL
- Pk:100 µl
MA3-926 detects sodium/calcium exchanger from human, canine, rabbit, mouse, guinea pig and rat kidney and cardiac tissues. MA3-926 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a 120 kDa protein representing the sodium/calcium exchanger from guinea pig cardiac extract. The bands seen at 70 kDa and 160 kDa represent a proteolytic fragment and non-reduced exchanger respectively. MA3-926 is not recommended for Western blot procedures of rat tissues. Immunohistochemical staining of the sodium/calcium exchanger in rat heart with MA3-926 results in staining of the plasma membrane and intense staining of cardiac T-tubular membrane. The MA3-926 antigen is purified canine cardiac sodium/calcium exchanger. This antibody recognizes an epitope between amino acids 371-525, which is on the intracellular side of the plasma membrane. The sodium/calcium exchanger of cardiac sarcolemma rapidly transports calcium during excitation-contraction coupling and is the dominant myocardial calcium efflux mechanism. The sodium/calcium exchanger uses the transmembrane sodium gradient to catalyze countertransport of calcium against its electrochemical gradient in a 3 sodium : 1 calcium exchange. Sodium/calcium exchange activity is present in excitable cells and in non-excitable cells.
Type: Primary
Antigen: SLC
Clonality: Monoclonal
Clone: C2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Canine, Guinea Pig, Rabbit, Rat, Mouse