Specifications
- Antibody type:Primary
- Antigen symbol:GCPII/PSMA
- Clonality:Monoclonal
- Clone:GCP-05
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:2346
- Antigen synonyms:glutamate carboxylase II|glutamate carboxypeptidase II|mGCP|pteroylpoly-gamma-glutamate carboxypeptidase|membrane glutamate carboxypeptidase|GCP2|FOLH|PSM|NAALAdase|GCPII|NAALAD1|N-acetylated alpha-linked acidic dipeptidase 1|PSMA|folylpoly-gamma-glutamate carboxypeptidase|N-acetylated-alpha-linked acidic dipeptidase I|prostate specific membrane antigen variant F|glutamate carboxypeptidase 2|FGCP|NAALADase I|GIG27|cell growth-inhibiting gene 27 protein
- Storage buffer:PBS, 0.2% BSA, 15mM sodium azide
- Storage temperature:4° C, store in dark, DO NOT FREEZE!
- Shipping temperature:Ice
- Immunogen:amino acids 44-750 of human GCPII
- Purification:purified
- Size:100 ug
- Pk:100 µG
Specifications
About this item
This antibody recognizes an extracellular domain, preferentially in native form, of glutamate carboxypeptidase II. This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants encoding several different isoforms. [provided by RefSeq, Jul 2010]
Type: Primary
Antigen: GCPII/PSMA
Clonality: Monoclonal
Clone: GCP-05
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human