Anti-GLP1R Rabbit Polyclonal Antibody
NOVUNLS1205
- Antibody type:Primary
- Antigen name:Glucagon-like peptide 1 receptor
- Antigen symbol:GLP1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:2740
- Antigen synonyms:GLP-1-R|GLP-1|Glucagon-like peptide-1 receptor|GLP-1 receptor|GLP-1R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
The GLP-1R Antibody from Novus Biologicals is a rabbit polyclonal antibody to GLP-1R. This antibody reacts with human, mouse. The GLP-1R Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 10 to 20 µg/ml
Type: Primary
Antigen: GLP-1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse