Anti-COL1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The Collagen I alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Collagen I alpha 1. This antibody reacts with human, mouse. The Collagen I alpha 1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: Collagen I alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-77458SS
- NOVUNBP1-77458
- Antigen symbol:
- COL1
- COL1
- Antigen name:
- Collagen I
- Collagen I
- Antigen synonyms:
- Collagen 1|Type I Collagen|COL1A
- Collagen 1|Type I Collagen|COL1A
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1277
- 1277
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together