Order Entry
Ireland
ContactUsLinkComponent
Anti-ABCA1 Rat Monoclonal Antibody [clone: 3A1.891.3]
Anti-ABCA1 Rat Monoclonal Antibody [clone: 3A1.891.3]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-ABCA1 Rat Monoclonal Antibody [clone: 3A1.891.3]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB400-164SS
  • NOVUNB400-164
  • Antigen symbol:
  • ABCA1
  • ABCA1
  • Antigen name:
  • ATP Binding Cassette, sub-family A, member 1
  • ATP Binding Cassette, sub-family A, member 1
  • Antigen synonyms:
  • ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
  • ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 3A1.891.3
  • 3A1.891.3
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19
  • 19
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together