Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB400-159SS
- NOVUNB400-159
- Antigen symbol:
- MSR1
- MSR1
- Antigen name:
- Macrophage Scavenger Receptor 1
- Macrophage Scavenger Receptor 1
- Antigen synonyms:
- MRS-A|Scvr|SR-AII|Scara1|MSR|MSR-A|CD204|SR-AI|MSR1
- MRS-A|Scvr|SR-AII|Scara1|MSR|MSR-A|CD204|SR-AI|MSR1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4481
- 4481
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together