Order Entry
Ireland
ContactUsLinkComponent
Anti-NES Mouse Monoclonal Antibody (DyLight® 680) [clone: 10C2]
Anti-NES Mouse Monoclonal Antibody (DyLight® 680) [clone: 10C2]
Catalog # NOVUNB300-266FR
Supplier:  Novus Biologicals
Anti-NES Mouse Monoclonal Antibody (DyLight® 680) [clone: 10C2]
Catalog # NOVUNB300-266FR
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nestin
  • Antigen symbol:
    NES
  • Clonality:
    Monoclonal
  • Clone:
    10C2
  • Conjugation:
    DyLight® 680
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    10763
  • Antigen synonyms:
    Nbla00170
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: NES (nestin)
Clonality: monoclonal
Clone: 10C2
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human