Anti-FXR2 Mouse Monoclonal Antibody (Biotin) [clone: 1G2]
NOVUNB300-259B
- Antibody type:Primary
- Antigen name:FMR1 autosomal homolog 2
- Antigen symbol:FXR2
- Clonality:Monoclonal
- Clone:1G2
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:9513
- Antigen synonyms:FMR1L2|FXR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
The FXR2 Antibody (1G2) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to FXR2. This antibody reacts with human, mouse. The FXR2 Antibody (1G2) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 20 ?g/ml
Type: Primary
Antigen: FXR2
Clonality: Monoclonal
Clone: 1G2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse