Anti-BEST1 Mouse monoclonal antibody unconjugated [clone: E6-6]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: BEST1 (bestrophin 1)
Clonality: monoclonal
Clone: E6-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- NOVUNB300-164SS
- NOVUNB300-164
- Antigen symbol:
- BEST
- BEST
- Antigen name:
- Bestrophin 1
- Bestrophin 1
- Antigen synonyms:
- BMD|TU15B|RP50|vitelliform macular dystrophy protein 2|Best disease|bestrophin-1|VMD2|ARB
- BMD|TU15B|RP50|vitelliform macular dystrophy protein 2|Best disease|bestrophin-1|VMD2|ARB
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- E6-6
- E6-6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7439
- 7439
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together