Anti-Neurokinin 1 Receptor Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB300-119C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Tachykinin Receptor 1
- Antigen symbol:TACR1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:6869
- Antigen synonyms:NK1R|NKIR|TAC1R|SPR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Neurokinin 1 Receptor
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat