Order Entry
Ireland
ContactUsLinkComponent
 
Anti-OPA1 Rabbit Polyclonal Antibody
 
undefined
Anti-OPA1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB110-55290SS
  • NOVUNB110-55290
  • Antigen symbol:
  • OPA1
  • OPA1
  • Antigen name:
  • Optic Atrophy 1
  • Optic Atrophy 1
  • Antigen synonyms:
  • optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
  • optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4976
  • 4976
  • Western blot:
  • Yes
  • Yes
  • Storage buffer:
  • PBS
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together