Order Entry
Ireland
ContactUsLinkComponent
 
Anti-TLR2 Rabbit Polyclonal Antibody
  NOVUNB100-56720
undefined
Anti-TLR2 Rabbit Polyclonal Antibody
  NOVUNB100-56720

 

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 2
  • Antigen symbol:
    TLR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    7097
  • Antigen synonyms:
    CD282|TIL4|Toll/interleukin-1 receptor-like protein 4
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

The TLR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR2. This antibody reacts with human, mouse. The TLR2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse