Anti-IRE1 alpha Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-2324F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:endoplasmic reticulum to nucleus signaling 1
- Antigen symbol:ERN1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:2081
- Antigen synonyms:hIRE1p|IRE1P|IRE1a|IRE1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: IRE1 alpha
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat