Anti-Tafazzin/TAZ Rabbit Polyclonal Antibody
Catalog # BSBTPA2135
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Tafazzin
- Antigen symbol:TAZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CMD3A|EFE|dilated 3A (X-linked) endocardial fibroelastosis 2 Barth syndrome) |EFE2|XAP-2 |OTTHUMP00000061673|tafazzin|TAZ|HGNC:11577|G4.5|dilated 3A (X-linked)|FLJ27390|XX-FW83563B9.3|tafazzin (cardiomyopathy|BTHS|LVNCX|cardiomyopathy|Taz1
- Amino acid number:162 - 178
- Sequence:MDFILEKLNHGDWVHIF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Tafazzin (162-178aa MDFILEKLNHGDWVHIF), identical to the related mouse and rat sequences
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Tafazzin(TAZ) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Tafazzin/TAZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat