Anti-IL2RA Rabbit Polyclonal Antibody
Catalog # BSBTPA1747
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 2 Receptor, Alpha
- Antigen symbol:IL2RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
- Amino acid number:39 - 57
- Sequence:MAYKEGTMLNCECKRGFRR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human IL2RA (39-57aa MAYKEGTMLNCECKRGFRR).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-2 receptor subunit alpha (IL2RA) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL2RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human