Order Entry
Ireland
ContactUsLinkComponent
Anti-LXR alpha Rabbit Polyclonal Antibody
Anti-LXR alpha Rabbit Polyclonal Antibody
  BSBTPA1246
undefined
Anti-LXR alpha Rabbit Polyclonal Antibody
  BSBTPA1246

 

  • Antibody type:
    Primary
  • Antigen name:
    Liver X receptor alpha
  • Antigen symbol:
    LXRA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    LXRA|LXR-a|RLD-1
  • Amino acid number:
    375 to 398
  • UniProtKB:
    Q13133
  • Sequence:
    QVERLQHTYVEALHAYVSINHPHD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human LXR alpha (375-398aa QVERLQHTYVEALHAYVSINHPHD), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Oxysterols receptor LXR-alpha(NR1H3) detection. Tested with WB in Human;Mouse;Rat.

LXRA is a tissue-specific cofactor that permits RXRA to function as a potent 9cRA receptor with a distinct target gene specificity. It specifically interacts with RXRA in vivo to form a functional heterodimer in which RXRA is the ligand-binding subunit. Additionally, LXR activity is critical for physiologic lipid metabolism and transport. LXRs are endogenous inhibitors of atherogenesis and are targets for therapeutic intervention in cardiovascular disease. Furthermore, LXRs and their ligands are negative regulators of macrophage inflammatory gene expression. LXR is also found that as a transcriptional switch that integrates hepatic glucose metabolism and fatty acid synthesis. The LXR-IDOL-LDLR axis defines a complementary pathway to sterol response element-binding proteins for sterol regulation of cholesterol uptake.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: LXR alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat