Anti-O-GlcNAc Mouse Monoclonal Antibody [clone: 1F5.D6]
Catalog # PIEAMA1040
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:O-linked N-acetylglucosamine
- Antigen symbol:O-GlcNAc
- Clonality:Monoclonal
- Clone:1F5.D6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:8473
- Antigen synonyms:O-GlcNAc transferase p110 subunit|O-GlcNAc transferase subunit p110|O-linked N-acetylglucosamine transferase 110 kDa subunit|O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase)|O-linked GlcNAc transferase
- Storage buffer:PBS, 1mg/ml BSA, 0.05% sodium azide
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:SANM-conjugated linear peptide corresponding to human O-GluNAc.
- Purification:Protein A
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MA1-040 has been successfully used in Western blot and immunoprecipitation applications with human samples. In Western blot, MA1-040 detects proteins with O-GlcNAc post-translational modification, and therefore will detect multiple specific bands. O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.
Type: Primary
Antigen: O-GlcNAc
Clonality: Monoclonal
Clone: 1F5.D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human