Anti-CDC25C Mouse Monoclonal Antibody [clone: IMD-25]
Catalog # BSBTMA1018
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 25C
- Antigen symbol:CDC25C
- Clonality:Monoclonal
- Clone:IMD-25
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:MPI1B|MPIP3|PPP1R60|CDC25M1|M-phase inducer phosphatase 3|EC 3.1.3.48
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human Cdc25c.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CDC25C
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human