Order Entry
Ireland
ContactUsLinkComponent
Anti-BTBD17 Rabbit Monoclonal Antibody [clone: EPR9593(B)]
Anti-BTBD17 Rabbit Monoclonal Antibody [clone: EPR9593(B)]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-BTBD17 Rabbit Monoclonal Antibody [clone: EPR9593(B)]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB138507-100
  • ABCAAB138507-1ML
  • Antigen symbol:
  • BTBD17
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together