Order Entry
Ireland
ContactUsLinkComponent
Anti-CBS Rabbit Monoclonal Antibody [clone: EPR8578]
Anti-CBS Rabbit Monoclonal Antibody [clone: EPR8578]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-CBS Rabbit Monoclonal Antibody [clone: EPR8578]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB131155-100
  • ABCAAB131155-1ML
  • Antigen symbol:
  • CBS
  • Antigen name:
  • cystathionine-beta-synthase
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR8578
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together