Order Entry
Ireland
ContactUsLinkComponent
Anti-NOL3 Rabbit Polyclonal Antibody
Anti-NOL3 Rabbit Polyclonal Antibody
Catalog # ABCAAB126238-50
Supplier:  Abcam
undefined
Anti-NOL3 Rabbit Polyclonal Antibody
Catalog # ABCAAB126238-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleolar protein 3
  • Antigen symbol:
    NOL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NOL3 (nucleolar protein 3 (apoptosis repressor with CARD domain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human