Order Entry
Ireland
ContactUsLinkComponent
Anti-RSPH4A Rabbit Polyclonal Antibody
Anti-RSPH4A Rabbit Polyclonal Antibody
  ABCAAB122159-100
 :  Abcam
Anti-RSPH4A Rabbit Polyclonal Antibody
  ABCAAB122159-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    radial spoke head 4 homolog A (Chlamydomonas)
  • Antigen symbol:
    RSPH4A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RSPH4A (radial spoke head 4 homolog A (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human