Order Entry
Ireland
ContactUsLinkComponent
Anti-HSPA5 Mouse Monoclonal Antibody (DyLight® 488) [clone: 10C3]
Anti-HSPA5 Mouse Monoclonal Antibody (DyLight® 488) [clone: 10C3]
Catalog # ABCAAB115638-50
Supplier:  Abcam
Anti-HSPA5 Mouse Monoclonal Antibody (DyLight® 488) [clone: 10C3]
Catalog # ABCAAB115638-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 70kDa protein 5
  • Antigen symbol:
    HSPA5
  • Clonality:
    Monoclonal
  • Clone:
    10C3
  • Conjugation:
    DyLight® 488
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: monoclonal
Clone: 10C3
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat