Anti-ATPB Mouse Monoclonal Antibody [clone: 7E3F2]
Catalog # ABCAAB110280-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:ATP Synthase beta subunit
- Antigen symbol:ATPB
- Clonality:Monoclonal
- Clone:7E3F2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ATPB
Clonality: monoclonal
Clone: 7E3F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: