Anti-Cre recombinase Mouse monoclonal antibody unconjugated [clone: DF6]
ABCAAB101071-100
: Abcam
- Antibody type:Primary
- Antigen name:Cre Recombinase
- Clonality:Monoclonal
- Clone:DF6
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cre recombinase
Clonality: monoclonal
Clone: DF6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: