Order Entry
Ireland
ContactUsLinkComponent
Anti-beta Catenin (phospho S45 + T41) Rabbit Monoclonal Antibody [clone: EP1905Y]
Anti-beta Catenin (phospho S45 + T41) Rabbit Monoclonal Antibody [clone: EP1905Y]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-beta Catenin (phospho S45 + T41) Rabbit Monoclonal Antibody [clone: EP1905Y]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB81305-20UL
  • ABCAAB81305-100
  • ABCAAB81305-1ML
  • Antigen symbol:
  • CTNNB1
  • Antigen name:
  • catenin (cadherin-associated Protein), beta 1, 88kDa
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EP1905Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together