Order Entry
Ireland
ContactUsLinkComponent
Anti-Pea3 Mouse Monoclonal Antibody [clone: 1A2G3+4B11E5+7D2E5]
Anti-Pea3 Mouse Monoclonal Antibody [clone: 1A2G3+4B11E5+7D2E5]
Catalog # ABCAAB70425-100
Supplier:  Abcam
Anti-Pea3 Mouse Monoclonal Antibody [clone: 1A2G3+4B11E5+7D2E5]
Catalog # ABCAAB70425-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Pea3
  • Clonality:
    Monoclonal
  • Clone:
    1A2G3+4B11E5+7D2E5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Pea3
Clonality: monoclonal
Clone: 1A2G3+4B11E5+7D2E5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human