Order Entry
Ireland
ContactUsLinkComponent
Anti-HSV1+ HSV2 ICP27 Mouse Monoclonal Antibody [clone: H1113]
Anti-HSV1+ HSV2 ICP27 Mouse Monoclonal Antibody [clone: H1113]
Catalog # ABCAAB53480-250
Supplier:  Abcam
Anti-HSV1+ HSV2 ICP27 Mouse Monoclonal Antibody [clone: H1113]
Catalog # ABCAAB53480-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSV1+ HSV2 ICP27
  • Clonality:
    Monoclonal
  • Clone:
    H1113
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HSV1+ HSV2 ICP27
Clonality: monoclonal
Clone: H1113
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: