Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PP2A alpha + beta Rabbit Monoclonal Antibody [clone: Y119]
 :  Abcam
New Product
 
undefined
Anti-PP2A alpha + beta Rabbit Monoclonal Antibody [clone: Y119]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB32141-1ML
  • ABCAAB32141-100
  • Antigen symbol:
  • PP2A alpha and beta
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • Y119
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together